Anti-EXT1 Rabbit polyclonal antibody

Supplier: Biorbyt
ORB48367-100
BIRBORB48367-100EA 0 EUR
BIRBORB48367-100
Anti-EXT1 Rabbit polyclonal antibody
Antibodies
Type: Primary
Antigen: EXT1 (exostosin glycosyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR