Anti-F12 Rabbit polyclonal antibody

Supplier: Biorbyt
BIRBORB48369-100EA 0 EUR
BIRBORB48369-100
Anti-F12 Rabbit polyclonal antibody
Antibodies
Type: Primary
Antigen: F12 (coagulation factor XII (Hageman factor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR