Anti-FCER1A Rabbit polyclonal antibody

Supplier: Biorbyt
BIRBORB48375-100EA 0 EUR
BIRBORB48375-100
Anti-FCER1A Rabbit polyclonal antibody
Antibodies
Type: Primary
Antigen: FCER1A (Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR