Anti-PRDX6 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB48505-100
BIRBORB48505-100EA 0 EUR
BIRBORB48505-100
Anti-PRDX6 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PRDX6 (peroxiredoxin 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR