Anti-TFPI Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB48555-100EA 0 EUR
BIRBORB48555-100
Anti-TFPI Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TFPI (tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR