Anti-TNFAIP3 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB48562-100
BIRBORB48562-100EA 0 EUR
BIRBORB48562-100
Anti-TNFAIP3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TNFAIP3 (tumor necrosis factor, alpha-induced protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR