Anti-KAT8 Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB48587-100EA 0 EUR
BIRBORB48587-100
Anti-KAT8 Mouse Monoclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: KAT8 (K(lysine) acetyltransferase 8)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR