Anti-PCT Chicken Polyclonal Antibody

Supplier: Biorbyt
BIRBORB48634-200EA 0 EUR
BIRBORB48634-200
Anti-PCT Chicken Polyclonal Antibody
Antibodies
ELISA: 1:5000-10000

Type: Primary
Antigen: PCT
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR