Anti-CNP Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB48654-200
BIRBORB48654-200EA 0 EUR
BIRBORB48654-200
Anti-CNP Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CNP (2',3'-cyclic nucleotide 3' phosphodiesterase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR