Anti-CARTPT Rabbit polyclonal antibody

Supplier: Biorbyt
ORB48723-200
BIRBORB48723-200EA 0 EUR
BIRBORB48723-200
Anti-CARTPT Rabbit polyclonal antibody
Antibodies
Type: Primary
Antigen: CARTPT (CART prepropeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR