Anti-APLNR Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB48742-200
BIRBORB48742-200EA 0 EUR
BIRBORB48742-200
Anti-APLNR Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: APLNR (apelin receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR