Anti-CCK Rabbit polyclonal antibody

Supplier: Biorbyt
ORB48755-200
BIRBORB48755-200EA 0 EUR
BIRBORB48755-200
Anti-CCK Rabbit polyclonal antibody
Antibodies
Type: Primary
Antigen: CCK (cholecystokinin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR