Anti-MSH alpha Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB48761-200EA 0 EUR
BIRBORB48761-200
Anti-MSH alpha Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MSH alpha
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR