Anti-Fibronectin Rabbit polyclonal antibody

Supplier: Biorbyt
BIRBORB48785-200EA 0 EUR
BIRBORB48785-200
Anti-Fibronectin Rabbit polyclonal antibody
Antibodies
Type: Primary
Antigen: Fibronectin (fibronectin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR