Anti-IL12 Rat Monoclonal Antibody

Supplier: Biorbyt
BIRBORB48795-500EA 0 EUR
BIRBORB48795-500
Anti-IL12 Rat Monoclonal Antibody
Antibodies
Type: Primary
Antigen: IL12
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG (Mouse)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR