Anti-MRSA Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB48867-200EA 0 EUR
BIRBORB48867-200
Anti-MRSA Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: MRSA
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG (Mouse)
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR