Anti-LEP Rabbit Monoclonal Antibody

Supplier: Biorbyt
BIRBORB48877-200EA 0 EUR
BIRBORB48877-200
Anti-LEP Rabbit Monoclonal Antibody
Antibodies
Type: Primary
Antigen: LEP (leptin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR