Anti-GHRH Rabbit monoclonal antibody

Supplier: Biorbyt
ORB48879-200
BIRBORB48879-200EA 0 EUR
BIRBORB48879-200
Anti-GHRH Rabbit monoclonal antibody
Antibodies
Type: Primary
Antigen: GHRH (growth hormone releasing hormone)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR