Anti-CD79 Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB48903-1000EA 0 EUR
BIRBORB48903-1000 BIRBORB48903-200
Anti-CD79 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: CD79
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR