Anti-CKB Mouse monoclonal antibody

Supplier: Biorbyt
ORB48904-200
BIRBORB48904-200EA 0 EUR
BIRBORB48904-200
Anti-CKB Mouse monoclonal antibody
Antibodies
Type: Primary
Antigen: CKB (creatine kinase, brain)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR