Anti-EEF1A1 Rabbit polyclonal antibody

Supplier: Biorbyt
BIRBORB48946-200EA 0 EUR
BIRBORB48946-200 BIRBORB48946-100
Anti-EEF1A1 Rabbit polyclonal antibody
Antibodies
Type: Primary
Antigen: EEF1A1 (eukaryotic translation elongation factor 1 alpha 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR