Anti-RAN Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB48948-100 ORB48948-200
BIRBORB48948-100EA 0 EUR
BIRBORB48948-100 BIRBORB48948-200
Anti-RAN Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RAN (RAN, member RAS oncogene family)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR