Anti-RHOC Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB48980-100 ORB48980-200
BIRBORB48980-100EA 0 EUR
BIRBORB48980-100 BIRBORB48980-200
Anti-RHOC Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RHOC (ras homolog family member C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR