Anti-ACTR3 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB48982-100EA 0 EUR
BIRBORB48982-100 BIRBORB48982-200
Anti-ACTR3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ACTR3 (ARP3 actin-related protein 3 homolog (yeast))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR