Anti-E2F8 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB49030-100EA 0 EUR
BIRBORB49030-100
Anti-E2F8 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: E2F8 (E2F transcription factor 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR