Anti-ACTN1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB49057-200EA 0 EUR
BIRBORB49057-200 BIRBORB49057-100
Anti-ACTN1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1500: 12000; Immunochemistry: 150: 1200

Type: Primary
Antigen: ACTN1 (actinin, alpha 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR