Anti-HNRNPA2B1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB49059-100 ORB49059-200
BIRBORB49059-100EA 0 EUR
BIRBORB49059-100 BIRBORB49059-200
Anti-HNRNPA2B1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HNRNPA2B1 (heterogeneous nuclear ribonucleoprotein A2/B1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR