Anti-PRKAR1A Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB49079-100EA 0 EUR
BIRBORB49079-100 BIRBORB49079-200
Anti-PRKAR1A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PRKAR1A (protein kinase, cAMP-dependent, regulatory, type I, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR