Anti-RAB5A Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB49080-100EA 0 EUR
BIRBORB49080-100 BIRBORB49080-200
Anti-RAB5A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RAB5A (RAB5A, member RAS oncogene family)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR