Anti-TARDBP Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB49083-100 ORB49083-200
BIRBORB49083-100EA 0 EUR
BIRBORB49083-100 BIRBORB49083-200
Anti-TARDBP Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TARDBP (TAR DNA binding protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR