Anti-MMP1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB49093-100EA 0 EUR
BIRBORB49093-100 BIRBORB49093-200
Anti-MMP1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MMP1 (matrix metallopeptidase 1 (interstitial collagenase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR