Anti-ECI1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB49114-100EA 0 EUR
BIRBORB49114-100 BIRBORB49114-200
Anti-ECI1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1500: 12000; Immunochemistry: 150: 1200

Type: Primary
Antigen: ECI1 (enoyl-CoA delta isomerase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR