Anti-STIP1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB49122-100 ORB49122-200
BIRBORB49122-100EA 0 EUR
BIRBORB49122-100 BIRBORB49122-200
Anti-STIP1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: STIP1 (stress-induced phosphoprotein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR