Anti-CGA Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB49142-200EA 0 EUR
BIRBORB49142-200 BIRBORB49142-100
Anti-CGA Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CGA (glycoprotein hormones, alpha polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR