Anti-SIRT4 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Biorbyt
ORB66479-100
BIRBORB66479-100EA 0 EUR
BIRBORB66479-100
Anti-SIRT4 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Western Blot: 1-1000

Type: Primary
Antigen: SIRT4 (sirtuin 4)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR