Anti-APOA4 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Biorbyt
ORB66481-100
BIRBORB66481-100EA 0 EUR
BIRBORB66481-100
Anti-APOA4 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Western Blot: 1-1000

Type: Primary
Antigen: APOA4 (apolipoprotein A-IV)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR