Anti-KT3 Tag Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB66655-100EA 0 EUR
BIRBORB66655-100 BIRBORB66655-20
Anti-KT3 Tag Mouse Monoclonal Antibody
Antibodies
Western Blot: 1:3000-1:10000

Type: Primary
Antigen: KT3 Tag
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR