Anti-IL17A Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB66667-100EA 0 EUR
BIRBORB66667-100
Anti-IL17A Mouse Monoclonal Antibody
Antibodies
Western Blot: 1:1000-1:5000

Type: Primary
Antigen: IL17A (interleukin 17A)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR