Anti-GR1 Mouse Monoclonal Antibody [clone: 5B5]

Supplier: Biorbyt
ORB66674-100
BIRBORB66674-100EA 0 EUR
BIRBORB66674-100
Anti-GR1 Mouse Monoclonal Antibody [clone: 5B5]
Antibodies
Western Blot: 1:1000-1:5000

Type: Primary
Antigen: GR1
Clonality: monoclonal
Clone: 5B5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR