Anti-IL12B Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB66687-100EA 0 EUR
BIRBORB66687-100
Anti-IL12B Mouse Monoclonal Antibody
Antibodies
ELISA: 1:1000-1:10000

Type: Primary
Antigen: IL12B (interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocyte maturation factor 2, p40))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR