Anti-ETV6 Rabbit polyclonal antibody

Supplier: Biorbyt
ORB66730-100
BIRBORB66730-100EA 0 EUR
BIRBORB66730-100
Anti-ETV6 Rabbit polyclonal antibody
Antibodies
Western Blot: 1:1000-1:5000

Type: Primary
Antigen: ETV6 (ets variant 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR