Anti-HAT1 Rabbit polyclonal antibody

Supplier: Biorbyt
BIRBORB66733-100EA 0 EUR
BIRBORB66733-100
Anti-HAT1 Rabbit polyclonal antibody
Antibodies
Western Blot: 1:1000-1:5000

Type: Primary
Antigen: HAT1 (histone acetyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR