Anti-IL33 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB66787-100
BIRBORB66787-100EA 0 EUR
BIRBORB66787-100
Anti-IL33 Rabbit Polyclonal Antibody
Antibodies
ELISA: 1:1000-1:10000

Type: Primary
Antigen: IL33 (interleukin 33)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR