Anti-RPS6KA3 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB66843-100
BIRBORB66843-100EA 0 EUR
BIRBORB66843-100
Anti-RPS6KA3 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000-1:5000

Type: Primary
Antigen: RPS6KA3 (ribosomal protein S6 kinase, 90kDa, polypeptide 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR