Anti-ZHX2 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB66919-100
BIRBORB66919-100EA 0 EUR
BIRBORB66919-100
Anti-ZHX2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:1000

Type: Primary
Antigen: ZHX2 (zinc fingers and homeoboxes 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR