Anti-KDM1B Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB66973-100
BIRBORB66973-100EA 0 EUR
BIRBORB66973-100
Anti-KDM1B Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000-1:5000

Type: Primary
Antigen: KDM1B (lysine (K)-specific demethylase 1B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR