Anti-KDM3A Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB67000-100
BIRBORB67000-100EA 0 EUR
BIRBORB67000-100
Anti-KDM3A Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000-1:5000

Type: Primary
Antigen: KDM3A (lysine (K)-specific demethylase 3A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR