Anti-Gamma chain Rabbit polyclonal antibody

Supplier: Biorbyt
ORB100270-100 ORB100270-200
BIRBORB100270-100EA 0 EUR
BIRBORB100270-100 BIRBORB100270-200
Anti-Gamma chain Rabbit polyclonal antibody
Antibodies
Western Blot: 1:1000-5000; ELISA: 1:2000-10000; Immunochemistry: 1:200-1000 (based on 0.5 mg/ml)

Type: Primary
Antigen: Gamma chain
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR