Anti-Mammaglobin A Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB100310-200EA 0 EUR
BIRBORB100310-200 BIRBORB100310-100
Anti-Mammaglobin A Rabbit Polyclonal Antibody
Antibodies
ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: Mammaglobin A
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR