Anti-LRP8 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB100362-100 ORB100362-20 ORB100362-200 ORB100362-5
BIRBORB100362-100EA 0 EUR
BIRBORB100362-100 BIRBORB100362-20 BIRBORB100362-200 BIRBORB100362-5
Anti-LRP8 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: LRP8 (low density lipoprotein receptor-related protein 8, apolipoprotein e receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR